Learn More
Enzo Life Sciences PGE2 ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900001
Encompass_Preferred
Highly sensitive PGE2 ELISA kit for inflammation and eicosanoid research. The PGE2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 2. Commercially available since 1992, this kit is widely cited in peer-reviewed publications. The non-radioactive ready-to-use liquid color-coded reagents reduce errors. Alternative Name: Prostaglandin E2. Sensitivity: 13.4 pg/ml (range 39.1 - 2,500 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-092