Learn More
Enzo Life Sciences PGE2 high sensitivity ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI930001
Encompass_Preferred
Ultrasensitive PGE2 ELISA kit for inflammation and eicosanoid research. The PGE2 high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 1 hour. Alternative Name: Prostaglandin E2. Sensitivity: 8.26 pg/ml (range 7.8 - 1,000 pg/ml). Assay Time: Overnight + 1 hour. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-665