Learn More
Enzo Life Sciences PGF2alpha ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901069
Encompass_Preferred
The PGF2α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. Broad dynamic range is suitable for a wide variety of sample types. Ready-to-use liquid color-coded reagents are safe, non-radioactive, and reduce laboratory errors. Alternative Name: Prostaglandin F2alpha. Sensitivity: 6.71 pg/ml (range 3.05 - 50,000 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: DxS Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-222