Learn More
Enzo Life Sciences PGF2alpha ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900069
Encompass_Preferred
The PGF2α EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. Broad dynamic range is suitable for a wide variety of sample types. Ready-to-use liquid color-coded reagents are safe, non-radioactive, and reduce laboratory errors. Alternative name: Prostaglandin F2alpha. Assay Time: <3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
Catalog No. 50-200-6834