Learn More
Enzo Life Sciences PGF2alpha high sensitivity ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI930069
Encompass_Preferred
Most sensitive PGF2α ELISA kit available for prostaglandin research. The PGF2α high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 3 hours. Absorbance is read at 405 nm. Simply the most sensitive quantification fo PGF2α commercially available in an enzyme immunoassay. Assay more samples without extraction. Alternative name: Prostaglandin F2alpha. Sensitivity: 0.98 pg/ml (range 1.95 - 2,000 pg/ml). Assay Time: Overnight + 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
Catalog No. 50-200-7109