Learn More
Enzo Life Sciences Prostaglandin F2alpha . tromethamine (10mg). CAS: 38562-01-5
SDP
Catalog No. 502011417
Encompass_Preferred
A major uterine luteolytic prostaglandin. Endogenous agonist of the prostanoid FP receptor (Kd=1.0 nM). Inhibits differentiation of 3T3-L1 preadipocytes. The product is not sterile. Alternative name: PGF2alpha . THAM. Purity: ≥99% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1417 Supplier Enzo Life Sciences Supplier No. BMLPG0090010
A major uterine luteolytic prostaglandin. Endogenous agonist of the prostanoid FP receptor (Kd=1.0 nM). Inhibits differentiation of 3T3-L1 preadipocytes. The product is not sterile. Alternative name: PGF2alpha . THAM. Purity: ≥99% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.