Learn More
Arbor Assays PGFM ELISA Kit (5 Plate)
SDP
Supplier:  Arbor Assays K022H5
Encompass_Preferred
The DetectX 13,14,Dihydro-15-keto-Prostaglandin F2α (PGFM) ELSIA Kit measures PGFM present in non-invasively collected fecal and urine samples, as well as plasma, serum and TCM samples.
Catalog No. 50-227-8420