Learn More
OriGene pGFP-A-shAAV shRNA Vector | 5ug
SDP
Catalog No. NC1869445
Encompass_Preferred
The pGFP-A-shAAV vector was created with an integrated turboGFP element to readily verify transfection efficiency. It incorporates both a ampicillin and puromycin resistance elements for greater selection capabilities. The AAV2-backbone vector enables the helper-free packaging method to produce AAV particles.
Catalog No. NC1869445 Supplier OriGene Supplier No. TR30034
The pGFP-A-shAAV vector was created with an integrated turboGFP element to readily verify transfection efficiency. It incorporates both a ampicillin and puromycin resistance elements for greater selection capabilities. The AAV2-backbone vector enables the helper-free packaging method to produce AAV particles.