Learn More
OriGene pGFP-C-shLenti shRNA Vector, 5 ug
SDP
Catalog No. NC1766994
Encompass_Preferred
pGFP-C-shLenti vector is a third generation Lentivector which requires the viral components carried in other vectors to produce viral particles. There are three major functional elements within the 5LTR and 3LTR: a shRNA expression cassette driven by an U6 promoter, a puromycin resistant gene driven by a SV40 promoter and a tGFP gene driven by a CMV promoter. All of them can be packaged to viral particles and transduced to many cell lines. The bacterial selection marker for the vector is Chloramphenicol.
Catalog No. NC1766994 Supplier OriGene Supplier No. TR30023
pGFP-C-shLenti vector is a third generation Lentivector which requires the viral components carried in other vectors to produce viral particles. There are three major functional elements within the 5LTR and 3LTR: a shRNA expression cassette driven by an U6 promoter, a puromycin resistant gene driven by a SV40 promoter and a tGFP gene driven by a CMV promoter. All of them can be packaged to viral particles and transduced to many cell lines. The bacterial selection marker for the vector is Chloramphenicol.