Learn More
Perkin Elmer US LLC PG-H2 Plus Hydrogen Generator, 500 mL/min
Catalog No. 502104896
Encompass_Preferred
Recommended for GC detector gas applications and GC, GC/MS carrier gas applications. Designed with automatic cascading capability.
Catalog No. 50-210-4896 Supplier Perkin Elmer US LLC Supplier No. N9308580
Add to Cart
Add to Cart
Request a Quote
Recommended for GC detector gas applications and GC, GC/MS carrier gas applications. Designed with automatic cascading capability.