Learn More
Genscript Corporation PGI 5F2 mAb Mouse
Supplier:  Genscript Corporation V0560110
Encompass
Pepsinogens aspartic proteinases are classified into two groups pepsinogen I (PGI) and pepsinogen II (PGII) PGI and PGII are mainly secreted by gastric chief cells They are useful in diagnosing severe atrophic gastritis and stomach cancer
Catalog No. 50-000-57034