Learn More
Aviva Systems Biology Corp PGK1 Antibody - C-terminal region (ARP48141_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48141_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PGK1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human PGK1. Alternate names: MGC117307, MGC142128, MGC8947, MIG10, PGKAApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-59727