Learn More
Aviva Systems Biology Corp PGK1 Antibody - N-terminal region (ARP48140_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48140_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PGK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PGK1. Alternate names: MGC117307, MGC142128, MGC8947, MIG10, PGKAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-59726