Learn More
Cell Signaling Technology PGK1 Monoclonal Antibody, Rabbit, Endogenous, Unconjugated, 100 μL
SDP
Catalog No. 502732411
Encompass
This rabbit monoclonal antibody is designed to detect endogenous levels of total PGK1 protein. The antibody is produced by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of the human PGK1 protein. PGK1 acts as an essential enzyme in the glycolysis pathway, catalyzing the conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate, and is implicated in cancer cell proliferation and metabolic regulation.
  • Validated for western blotting applications
  • Reactivity in human, mouse, and rat samples
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and < 0.02% sodium azide
  • Targeted molecular weight of 43 kDa
Catalog No. 50-273-2411 Supplier Cell Signaling Technology Supplier No. 63536S
This rabbit monoclonal antibody is designed to detect endogenous levels of total PGK1 protein. The antibody is produced by immunizing animals with a synthetic peptide corresponding to residues near the carboxy terminus of the human PGK1 protein. PGK1 acts as an essential enzyme in the glycolysis pathway, catalyzing the conversion of 1,3-diphosphoglycerate to 3-phosphoglycerate, and is implicated in cancer cell proliferation and metabolic regulation.
  • Validated for western blotting applications
  • Reactivity in human, mouse, and rat samples
  • Supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and < 0.02% sodium azide
  • Targeted molecular weight of 43 kDa