Learn More
Medchemexpress LLC PGK1 Human HEK293 5ug
SDP
Catalog No. 5000196795
Encompass_Preferred
Recombinant full-length human phosphoglycerate kinase 1 produced in HEK293 cells and supplied as a C-terminal His-tagged protein (5 μg). The protein is purified to >95% and formulated in a buffered glycerol solution for stability, making it suitable for enzymatic characterization, binding assays, and biophysical analyses. Store frozen and avoid repeated freeze-thaw cycles for best activity.
  • High purity greater than 95% by reducing SDS-PAGE.
  • Expressed in human HEK293 cells for native post-translational modifications.
  • C-terminal 6xHis tag for straightforward purification and immobilization.
  • Supplied in buffered solution with glycerol for enhanced stability.
  • Suitable for enzymatic assays, binding studies, and SPR applications.
  • Store at -80°C and avoid repeated freeze-thaw cycles.
Catalog No. 50-001-96795 Supplier Medchemexpress LLC Supplier No. HYP710495UG
Recombinant full-length human phosphoglycerate kinase 1 produced in HEK293 cells and supplied as a C-terminal His-tagged protein (5 μg). The protein is purified to >95% and formulated in a buffered glycerol solution for stability, making it suitable for enzymatic characterization, binding assays, and biophysical analyses. Store frozen and avoid repeated freeze-thaw cycles for best activity.
  • High purity greater than 95% by reducing SDS-PAGE.
  • Expressed in human HEK293 cells for native post-translational modifications.
  • C-terminal 6xHis tag for straightforward purification and immobilization.
  • Supplied in buffered solution with glycerol for enhanced stability.
  • Suitable for enzymatic assays, binding studies, and SPR applications.
  • Store at -80°C and avoid repeated freeze-thaw cycles.