Learn More
Medchemexpress LLC PGK1 protein, mouse (sf9, His) | >95.0% | 5 UG
SDP
Catalog No. 5000293359
Encompass_Preferred
Recombinant full-length mouse phosphoglycerate kinase 1 (PGK1) expressed in Sf9 insect cells, supplied as a lyophilized sterile powder with a C-terminal His tag. The protein is formulated from 20 mM Tris, 500 mM NaCl, pH 7.4, provided with a datasheet, COA, and SDS, and is intended for research use.
  • Full-length mouse PGK1 (M1-V417) with C-terminal His tag.
  • Expressed in Sf9 insect cells for eukaryotic folding.
  • Lyophilized sterile powder formulation for long-term storage.
  • Purity >95.0% as determined by Tris-Bis PAGE; ~46 kDa.
  • Low endotoxin level (<1 EU/μg) and recommended storage at -20°C.
Catalog No. 50-002-93359 Supplier Medchemexpress LLC Supplier No. HYP746345UG
Recombinant full-length mouse phosphoglycerate kinase 1 (PGK1) expressed in Sf9 insect cells, supplied as a lyophilized sterile powder with a C-terminal His tag. The protein is formulated from 20 mM Tris, 500 mM NaCl, pH 7.4, provided with a datasheet, COA, and SDS, and is intended for research use.
  • Full-length mouse PGK1 (M1-V417) with C-terminal His tag.
  • Expressed in Sf9 insect cells for eukaryotic folding.
  • Lyophilized sterile powder formulation for long-term storage.
  • Purity >95.0% as determined by Tris-Bis PAGE; ~46 kDa.
  • Low endotoxin level (<1 EU/μg) and recommended storage at -20°C.