Learn More
Medchemexpress LLC HY-P0274A 1mg Medchemexpress, PGLa (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0274A1MG
Encompass_Preferred
Medchemexpress, HY-P0274A 1mg PGLa (TFA) CAS: PGLa TFA, a 21-residue peptide, is an antimicrobial peptide. PGLa TFA is a member of the magainin family of antibiotic peptides found in frog skin and its secretions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0522