Learn More
Aviva Systems Biology Corp PGLS Antibody - middle region (ARP58513_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58513_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PGLS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PGLS. Alternate names: 6PGLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62334