Learn More
Medchemexpress LLC HY-107380A 10mg Medchemexpress, PGlu-3-methyl-His-Pro-NH2 (TFA) CAS: Purity:>98%
SDP
Catalog No. 501968393
Encompass_Preferred
Medchemexpress, HY-107380A 10mg PGlu-3-methyl-His-Pro-NH2 (TFA) CAS: PGlu-3-methyl-His-Pro-NH2 TFA (A-42872 TFA), the modified thyrotropin-releasing hormone (TRH) peptide, enhances binding to pituitary TRH receptors and increases stimulation of thyroid-stimulating hormone (TSH) release from the pituitary. The in vitro permeability of PGlu-3-methyl-His-Pro-NH2 TFA through rat skin is increased in the presence of enhancers Ethanol and Cineole. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8393 Supplier Medchemexpress LLC Supplier No. HY107380A10MG
Medchemexpress, HY-107380A 10mg PGlu-3-methyl-His-Pro-NH2 (TFA) CAS: PGlu-3-methyl-His-Pro-NH2 TFA (A-42872 TFA), the modified thyrotropin-releasing hormone (TRH) peptide, enhances binding to pituitary TRH receptors and increases stimulation of thyroid-stimulating hormone (TSH) release from the pituitary. The in vitro permeability of PGlu-3-methyl-His-Pro-NH2 TFA through rat skin is increased in the presence of enhancers Ethanol and Cineole. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.