Learn More
AnaSpec PGLU-?-AMYLOID(3-42)
Supplier:  AnaSpec AS299071
Encompass
[Pyr3]-beta-Amyloid (3-42), Human[Pyr-FRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA] ; MW 4309.9; (1 mg) This is one of the predominant amyloid peptide structures deposited in human brain of Alzheimer’s disease and Down’s syndrome patients.
Catalog No. 50-824-567