Learn More
Medchemexpress LLC HY-P0022A 5mg Medchemexpress, pGlu-Pro-Arg-MNA (monoacetate) CAS:2070009-26-4 Purity:>98%
SDP
Catalog No. 501933996
Encompass_Preferred
Medchemexpress, HY-P0022A 5mg pGlu-Pro-Arg-MNA (monoacetate) CAS:2070009-26-4 pGlu-Pro-Arg-MNA monoacetate is a chromogenic substrate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3996 Supplier Medchemexpress LLC Supplier No. HYP0022A5MG
Medchemexpress, HY-P0022A 5mg pGlu-Pro-Arg-MNA (monoacetate) CAS:2070009-26-4 pGlu-Pro-Arg-MNA monoacetate is a chromogenic substrate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.