Learn More
Cayman Chemical [pGlu4]-Myelin Basic Protein Peptide (trifluoroacetate salt), Purity: ≥98%, 25 mg
SDP
Catalog No. 502942440
Encompass_Preferred
A peptide substrate for PKC; selectively phosphorylated by PKC over PKA and phosphorylase kinase
Catalog No. 50-294-2440 Supplier Cayman Chemical Supplier No. 4014925 MG
A peptide substrate for PKC; selectively phosphorylated by PKC over PKA and phosphorylase kinase