Learn More
Medchemexpress LLC PGLYRP4/PGRP-I beta 10ug | 10ug
SDP
Catalog No. 5000186288
Encompass_Preferred
PGLYRP4/PGRP-I beta protein is a model receptor that binds to the cell wall peptide peptidoglycan (PGN) in Gram-positive bacteria and exerts bactericidal activity by interfering with peptidoglycan biosynthesis Its antimicrobial properties extend to Gram-negative bacteria although it also binds to them PGLYRP4/PGRP-I beta Protein Human (HEK293 His) is the recombinant human-derived PGLYRP4/PGRP-I beta protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86288 Supplier Medchemexpress LLC Supplier No. HYP7713710UG
PGLYRP4/PGRP-I beta protein is a model receptor that binds to the cell wall peptide peptidoglycan (PGN) in Gram-positive bacteria and exerts bactericidal activity by interfering with peptidoglycan biosynthesis Its antimicrobial properties extend to Gram-negative bacteria although it also binds to them PGLYRP4/PGRP-I beta Protein Human (HEK293 His) is the recombinant human-derived PGLYRP4/PGRP-I beta protein expressed by HEK293 with C-His labeled tag