Learn More
Lonza Walkersville PBM-2 Preadipocyte Basal Medium-2
Supplier:  Lonza Walkersville PT8202
Encompass
PGM -2 Preadipocyte Growth Medium-2 BulletKit Medium provides both a proliferation medium and a differentiation medium guaranteed to induce differentiation into mature, functionally active adipocytes; Each bottle of the PGM-2 BulletKit Medium contains enough differentiation medium to fully differentiate 10 million preadipocytes [10 complete 96-well plates] into mature, functionally active adipocytes. The PGM-2 Medium is offered as a BulletKit Medium (catalog no. PT-8002) which includes both the basal medium and the necessary supplements for both proliferation and differentiation of preadipocytes. The basal medium (PGM-2 Basal Medium; catalog no. PT-8202) and the necessary supplements (PGM-2 SingleQuot Kit; catalog no. PT-9502) can also be purchased individually.
Catalog No. NC9409636