Learn More
Aviva Systems Biology Corp PGM1 Antibody - middle region (ARP56383_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56383_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PGM1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PGM1. Alternate names: GSD14ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-147