Learn More
Aviva Systems Biology Corp PGM3 Antibody - N-terminal region (ARP56755_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56755_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PGM3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PGM3. Alternate names: AGM1, DKFZp434B187, PAGM, PGM 3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61974