Learn More
Medchemexpress LLC HY-101143 100mg Medchemexpress, PGMI-004A CAS:1313738-90-7 Purity:>98%
SDP
Catalog No. 501932051
Encompass_Preferred
Medchemexpress, HY-101143 100mg PGMI-004A CAS:1313738-90-7 PGMI-004A is a potent phosphoglycerate mutase 1 ( PGAM1 ) inhibitor with an IC 50 of 13.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2051 Supplier Medchemexpress LLC Supplier No. HY101143100MG
Medchemexpress, HY-101143 100mg PGMI-004A CAS:1313738-90-7 PGMI-004A is a potent phosphoglycerate mutase 1 ( PGAM1 ) inhibitor with an IC 50 of 13.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.