Learn More
Invivogen PGN-BS peptidoglycan preparation, 5 mg
SDP
Supplier:  Invivogen TLRLPGNB3
Encompass
PGN-BS is a peptidoglycan (PGN) preparation from the Gram-positive Bacillus subtilis. PGN is a major component of the bacterial cell wall. It is mainly single-layered in Gram-negative bacteria (e.g. E. coli) and multi-layered in Gram-positive bacteria (e.g. B. subtilis). PGN is a large polymeric molecule made of glycan strands connected by short peptides. PGN structure varies among bacterial species at the level of sugar polymers, and of cross-linked amino-acids.
Catalog No. NC0664895