Learn More
Enzo Life Sciences PGP-4008 (2mg). CAS: 365565-02-2
SDP
Supplier:  Enzo Life Sciences ALX270290M002
Encompass_Preferred
Selective P-glycoprotein (Pgp) inhibitor. Effectively reverses Pgp mediated drug resistance in NCI/ADR cells, but has no effect on the drug sensitivities of MCF-7/VP cells that overexpress MRP1. Alternative name: N-(1-Benzyl-2,3-dihydro-1H-pyrrolo[2,3-b]quinolin-4-yl)-2-phenylacetamide. Formula: C26H23N3O. MW: 393.5. Purity: ≥93%. Solubility: Soluble in methanol, 100% ethanol (3mg/ml) or DMSO (20mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8471