Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pgp-4008 | 365565-02-2 | 99.3% | 1 ML
SDP

Catalog No. 5000234705
Encompass_Preferred

PGP-4008 is a small-molecule P-glycoprotein inhibitor provided as a 10 mM solution in DMSO (1 mL) for in-vitro research. It is used to study Pgp-mediated multidrug resistance and transport assays, with high purity and characterized physicochemical data.

  • Provided as 10 mM solution in DMSO, 1 mL
  • High purity: 99.26%
  • Molecular formula: C26H23N3O
  • Molecular weight: 393.48 g/mol
  • Solubility: DMSO soluble; ultrasonic and warming to 60°C recommended
  • Storage: in solvent -80°C (6 months); -20°C (1 month)

Catalog No. 50-002-34705 Supplier Medchemexpress LLC Supplier No. HY11982310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PGP-4008 is a small-molecule P-glycoprotein inhibitor provided as a 10 mM solution in DMSO (1 mL) for in-vitro research. It is used to study Pgp-mediated multidrug resistance and transport assays, with high purity and characterized physicochemical data.

  • Provided as 10 mM solution in DMSO, 1 mL
  • High purity: 99.26%
  • Molecular formula: C26H23N3O
  • Molecular weight: 393.48 g/mol
  • Solubility: DMSO soluble; ultrasonic and warming to 60°C recommended
  • Storage: in solvent -80°C (6 months); -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.