Learn More
Abcam PGP 4008, P-glycoprotein inhibitor, 10MG
Catalog No. 502705414
Encompass_Preferred
MW 393.5 Da, Purity >93%. Selective P-glycoprotein inhibitor. Reverses Pgp mediated drug resistance in NCI/ADR cells. Shows synergistic effects with doxorubicin (ab120629) in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5414 Supplier Abcam Supplier No. AB14519110MG
MW 393.5 Da, Purity >93%. Selective P-glycoprotein inhibitor. Reverses Pgp mediated drug resistance in NCI/ADR cells. Shows synergistic effects with doxorubicin (ab120629) in vivo. .
The product is subject to the following: Abcam Restricted Use Statement