Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7345MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: WB, IHC. Conjugate/Tag: Unconjugated. Clone: UCHL1/775. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Cerebellum. Cellular Staining: Cytoplasmic. Endoplasmic Reticulum membrane. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human UCHL1 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 20-30kDa.Chromosome: 4p13. Gene Symbol: UCHL1. Gene ID: 7345. Ensemble: ENSG00000154277. Uniprot: NP_004172.
Catalog No. 50-188-5265