Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5241MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: PR484. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: T47-D Cells or Breast Cancers. Cellular Staining: Nucleus. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Progesterone Receptor protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: PR-A (81kDa), PR-B (116kDa).Chromosome: 11q22.1. Gene Symbol: PGR. Gene ID: 5241. Ensemble: ENSG00000082175. Uniprot: NP_001189403.
Catalog No. 50-188-5051