Learn More
Aviva Systems Biology Corp PGR Antibody - N-terminal region (ARP38005_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PGR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PGR. Alternate names: NR3C3, PRApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-56484