Learn More
Aviva Systems Biology Corp Pgr Antibody - middle region (ARP36958_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36958_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pgr. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Pgr. Alternate names: 9930019P03Rik, NR3C3, PR, PR-A, PR-B, BB114106, ENSMUSG00000074510ApplicationWB. Species reactivity: Dog, Guinea Pig, Mouse, Rat
Catalog No. 50-284-552