Learn More
System Biosciences LLC pGreenFire1-ISRE (EF1α-neo) LentivectorpGreenFire1-ISRE (plasmid)+ EF1α-Neo= 10 µg
SDP
Catalog No. NC0891026
Encompass_Preferred
Study ISRE signal transduction by co-expressing dscGFP and luciferase in response to ISRE activity (includes EF1α-neo selection cassette). With SBI’s line of pGreenFire1 Pathway Reporters, you can monitor signal transduction in real time. These vectors leverage our reliable lentivector technology and save you time—our pre-built signal transduction pathway reporters come as ready-to-package lentivector plasmid and ready-to-transduce pre-packaged lentivirus*. The pGreenFire1-ISRE (EF1α-neo) Lentivector co-expresses a destabilized copepod GFP (dscGFP, 2-hour half-life) and luciferase from ISRE transcriptional response elements (TREs) paired with a minimal CMV promoter (mCMV). The mCMV promoter alone delivers negligible expression, but when downstream of ISRE-responsive transcriptional elements, drives expression of dscGFP and luciferase in response to ISRE activity. The result is the ability to quantitatively measure ISRE activity by fluorescence and luciferase activity.
Catalog No. NC0891026 Supplier System Biosciences LLC Supplier No. TR016PAN
Study ISRE signal transduction by co-expressing dscGFP and luciferase in response to ISRE activity (includes EF1α-neo selection cassette). With SBI’s line of pGreenFire1 Pathway Reporters, you can monitor signal transduction in real time. These vectors leverage our reliable lentivector technology and save you time—our pre-built signal transduction pathway reporters come as ready-to-package lentivector plasmid and ready-to-transduce pre-packaged lentivirus*. The pGreenFire1-ISRE (EF1α-neo) Lentivector co-expresses a destabilized copepod GFP (dscGFP, 2-hour half-life) and luciferase from ISRE transcriptional response elements (TREs) paired with a minimal CMV promoter (mCMV). The mCMV promoter alone delivers negligible expression, but when downstream of ISRE-responsive transcriptional elements, drives expression of dscGFP and luciferase in response to ISRE activity. The result is the ability to quantitatively measure ISRE activity by fluorescence and luciferase activity.