Learn More
System Biosciences LLC pGreenZeo-CMV Differentiation Reporter Positive ControlpGreenZeo-CMV Virus (positive control) = >2 x 10^6 IFUs
SDP
Catalog No. NC3500321
Encompass_Preferred
Run your pGreenZeo projects with confidence with the addition of this positive control that drives copGFP and zeomycin resistance with a CMV promoter. No need to make a positive control for your pGreenZeo projects—SBI’s already built one for you. No need to make a positive control for your pGreenZeo projects—SBI’s already built one for you. The GreenZeo cassette is driven by the strong, constitutive CMV promoter with the GFP and zeomycin marker co-expression mediated by a T2A element.
Catalog No. NC3500321 Supplier System Biosciences LLC Supplier No. SR501VA1
Run your pGreenZeo projects with confidence with the addition of this positive control that drives copGFP and zeomycin resistance with a CMV promoter. No need to make a positive control for your pGreenZeo projects—SBI’s already built one for you. No need to make a positive control for your pGreenZeo projects—SBI’s already built one for you. The GreenZeo cassette is driven by the strong, constitutive CMV promoter with the GFP and zeomycin marker co-expression mediated by a T2A element.