Learn More
System Biosciences LLC pGreenZeo-mCMV Differentiation Reporter Negative ControlpGreenZeo-mCMV Plasmid (negative control) =10 µg
SDP
Catalog No. NC3500252
Encompass_Preferred
Run your pGreenZeo projects with confidence with the addition of this negative control that drives dscGFP and zeomycin resistance with a minimal CMV promoter. No need to make a negative control for your pGreenZeo projects—SBI’s already built one for you. The GreenZeo cassette is driven by a minimal CMV promoter with the dscGFP (destabilized copGFP, 2-hour half-life) and zeomycin marker co-expression mediated by a T2A element. This configuration provides a control for background levels of GFP and zeomycin expression in the absence of enhancer elements, such as the ones used in our Pluripotency Reporters.
Catalog No. NC3500252 Supplier System Biosciences LLC Supplier No. SR500PA1
Run your pGreenZeo projects with confidence with the addition of this negative control that drives dscGFP and zeomycin resistance with a minimal CMV promoter. No need to make a negative control for your pGreenZeo projects—SBI’s already built one for you. The GreenZeo cassette is driven by a minimal CMV promoter with the dscGFP (destabilized copGFP, 2-hour half-life) and zeomycin marker co-expression mediated by a T2A element. This configuration provides a control for background levels of GFP and zeomycin expression in the absence of enhancer elements, such as the ones used in our Pluripotency Reporters.