Learn More
Medchemexpress LLC HY-112798 10mg Medchemexpress, PH-002 CAS:1311174-68-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11279810MG
Encompass_Preferred
Medchemexpress, HY-112798 10mg PH-002 CAS:1311174-68-1 PH-002 is an inhibitor of apolipoprotein (apo) E4 intramolecular domain interaction in neuronal cells that could rescue impairments of mitochondrial motility and neurite outgrowth. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9173