Learn More
Macherey-Nagel pH test paper Tritest pH 1-11, with three indicator zones
Catalog No. NC0344351
Encompass_Preferred
Tritest indicator paper has three different indicator zones on a single paper. The three zones guarantee optimal color differences and safe determination of in-between values.
Catalog No. NC0344351 Supplier Macherey-Nagel Supplier No. 90501
Tritest indicator paper has three different indicator zones on a single paper. The three zones guarantee optimal color differences and safe determination of in-between values.