Learn More
Andwin Scientific PH 10 BLUE BUFFER
SDP
Catalog No. NC0429524
Encompass_Preferred
The BDH pH reference standard buffer is a clear, colorless, odorless, liquid solution. This contains potassium tetroxalate dihydrate and deionized water. It is classified to have a 10.00 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.
Catalog No. NC0429524 Supplier Andwin Scientific Supplier No. BDH5072
The BDH pH reference standard buffer is a clear, colorless, odorless, liquid solution. This contains potassium tetroxalate dihydrate and deionized water. It is classified to have a 10.00 pH at 25.0 C. This product is packaged and sold in a 500 mL poly bottle.