Learn More
Andwin Scientific PH 10 BUFFER SOLUTION, 4L
SDP
Catalog No. NC0389127
Encompass_Preferred
Buffer Reference Standard^ pH 10.00 +- 0.01, Color Coded Blue, 4 L Cubitainer
Catalog No. NC0389127 Supplier Andwin Scientific Supplier No. BDH50764L
Buffer Reference Standard^ pH 10.00 +- 0.01, Color Coded Blue, 4 L Cubitainer