Learn More
HANNA INSTRUMENTS INC PH 10 BUFFER STANDARD 500ML
SDP
Catalog No. NC2306241
Encompass
HI7010L is a premium quality pH 10.01 calibration solution. Hanna calibration buffers have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution. Hanna’s line of calibration buffers have been specially formulated to have an expiration of 2 years from the date of manufacture for an unopened bottle.Color coded label for easy pH buffer value identificationAccuracy of +/- 0.01 pH @ 25°CTemperature chart of the actual pH value at various temperatures printed on each bottle
Catalog No. NC2306241 Supplier HANNA INSTRUMENTS INC Supplier No. HI7010L
HI7010L is a premium quality pH 10.01 calibration solution. Hanna calibration buffers have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution. Hanna’s line of calibration buffers have been specially formulated to have an expiration of 2 years from the date of manufacture for an unopened bottle.Color coded label for easy pH buffer value identificationAccuracy of +/- 0.01 pH @ 25°CTemperature chart of the actual pH value at various temperatures printed on each bottle