Learn More
Waters Corp Waters Corporation pH Buffer, pH 10, 1 Liter, NIST Traceable
Catalog No. 50465882 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pH 10 buffer is designed for calibration purposes, traceable to NIST SRMs, and is mercury-free. It is guaranteed stable for at least one year after receipt and includes a full certificate of analysis, suitable for applications requiring precise pH measurement, such as those related to EPA Method 150.1.
  • Liquid formulation
  • Storage at room temperature
  • Shelf life of 12 months
  • NIST traceable
  • Ambient shipping condition
  • 1 Liter volume
  • Calibration type
  • Meets EPA method 150.1
  • Mercury-free
  • Supplied with a full certificate of analysis
Catalog No. 50-465-882 Supplier Waters Corp Supplier No. 137
This pH 10 buffer is designed for calibration purposes, traceable to NIST SRMs, and is mercury-free. It is guaranteed stable for at least one year after receipt and includes a full certificate of analysis, suitable for applications requiring precise pH measurement, such as those related to EPA Method 150.1.
  • Liquid formulation
  • Storage at room temperature
  • Shelf life of 12 months
  • NIST traceable
  • Ambient shipping condition
  • 1 Liter volume
  • Calibration type
  • Meets EPA method 150.1
  • Mercury-free
  • Supplied with a full certificate of analysis