Learn More
Waters Corp Waters Buffer, pH 10, Pint, Liquid, NIST Traceable
Catalog No. 50465881 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This pH Buffer is directly traceable to NIST SRMs, mercury-free, guaranteed stable for at least one year after your receipt, and is supplied with a full certificate of analysis. It comes in a 1 Pint Bottle.
  • Liquid formulation
  • Room temperature storage
  • 12 months shelf life
  • NIST traceable
  • Ambient shipping condition
  • For calibration
  • EPA Method 150.1
  • 1 unit per package
Catalog No. 50-465-881 Supplier Waters Corp Supplier No. 135
This pH Buffer is directly traceable to NIST SRMs, mercury-free, guaranteed stable for at least one year after your receipt, and is supplied with a full certificate of analysis. It comes in a 1 Pint Bottle.
  • Liquid formulation
  • Room temperature storage
  • 12 months shelf life
  • NIST traceable
  • Ambient shipping condition
  • For calibration
  • EPA Method 150.1
  • 1 unit per package