Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions pH 11.00 Buffer Solution with Boric Acid, 20 L
SDP

Catalog No. NC3351099
Encompass_Preferred

Aqua Solutions pH 11.00 reference buffer is a high-purity solution designed for precise pH calibration and analytical procedures in scientific and laboratory settings. With a large 20L volume, it ensures an ample supply for routine applications, delivering reliable and consistent pH measurements for various chemical and biological processes.

  • pH value of 11.00
  • Boric acid concentration of 0.2 Molar
  • Volume of 20 L
  • Product type: Buffer solution
  • Form: Liquid solution
  • Applications in pH calibration and analytical chemistry
  • Reference grade purity implied by its designation

Catalog No. NC3351099 Supplier Aqua Solutions Supplier No. DC020185
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Aqua Solutions pH 11.00 reference buffer is a high-purity solution designed for precise pH calibration and analytical procedures in scientific and laboratory settings. With a large 20L volume, it ensures an ample supply for routine applications, delivering reliable and consistent pH measurements for various chemical and biological processes.

  • pH value of 11.00
  • Boric acid concentration of 0.2 Molar
  • Volume of 20 L
  • Product type: Buffer solution
  • Form: Liquid solution
  • Applications in pH calibration and analytical chemistry
  • Reference grade purity implied by its designation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.