Learn More
YMC America, Inc Ph 120A 5um 75x0.5mm 1/16
SDP
Catalog No. 5000878856
Encompass
Phenyl columns provide unique selectivity when compared to aliphatic straight chain reversed phase materials such as C18 C8 and C4 YMC-Pack Phenyl is a monomerically-bonded phenyl and is fully endcapped Typical applications include aromatic compounds phenols fullerenes sweeteners
Catalog No. 50-008-78856 Supplier YMC America, Inc Supplier No. PH12S05L5J0AU
Phenyl columns provide unique selectivity when compared to aliphatic straight chain reversed phase materials such as C18 C8 and C4 YMC-Pack Phenyl is a monomerically-bonded phenyl and is fully endcapped Typical applications include aromatic compounds phenols fullerenes sweeteners