Learn More
Andwin Scientific PH Red Reagent Solution 500ml
SDP
Catalog No. NC9888474
Encompass_Preferred
The BDH pH reference standard buffer is a clear, light red, odorless, liquid solution. This contains hydrochloric acid, potassium chloride, and deionized water. It is classified to have a 3.99-4.01 pH at 25.0C. This product is packaged and sold in a 500 mL poly bottle.
Catalog No. NC9888474 Supplier Andwin Scientific Supplier No. BDH5018
The BDH pH reference standard buffer is a clear, light red, odorless, liquid solution. This contains hydrochloric acid, potassium chloride, and deionized water. It is classified to have a 3.99-4.01 pH at 25.0C. This product is packaged and sold in a 500 mL poly bottle.