Learn More
Aviva Systems Biology Corp PH-4 Antibody - middle region (ARP57762_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57762_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PH-4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PH-4. Alternate names: P4H-TM, PH4, PH-4, PHD4, EGLN4, HIFPH4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62149