Learn More
Andwin Scientific PH 4 BUFFER, POUCH 4L
SDP
Catalog No. NC0389125
Encompass_Preferred
The BDH pH reference standard buffer is a clear, red, odorless, liquid solution. It is classified to have a 3.99-4.01 pH at 25.0 �C. This product is packaged and sold in a 4 L cubitainer.
Catalog No. NC0389125 Supplier Andwin Scientific Supplier No. BDH50224L
The BDH pH reference standard buffer is a clear, red, odorless, liquid solution. It is classified to have a 3.99-4.01 pH at 25.0 �C. This product is packaged and sold in a 4 L cubitainer.